Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nos
Entrez Gene ID
34495
Gene Name
Nitric oxide synthase
Gene Aliases
CG6713, DNOS, DNOS1, Dmel\CG6713, Dmel_CG6713, NOS, dNOS, dNos, drNOSoxy, iNOS, nNOS, nos
Chromosome Location
Chr.2L: 10804274 - 10837511 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011676
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032069.2 NP_001027240.1 4 - 5 1249 80
NM_001032072.2 NP_001027243.2 2 - 3 553 80
NM_078817.4 NP_523541.2 4 - 5 1249 80
GenBank mRNA BT010016.1 - 4 - 5 1248 80

View Full Product Details