Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Acox57D-d
- Entrez Gene ID
-
37446
- Gene Name
- acyl-Coenzyme A oxidase at 57D distal
- Gene Aliases
-
Acox57Dd, CG9709, D-acox, Dmel\CG9709, Dmel_CG9709, aco, acox
- Chromosome Location
-
Chr.2R: 21389405 - 21392439 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034629
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079079.4 |
NP_523803.1 |
2 - 3 |
404 |
63 |
GenBank mRNA |
AY070539.1 |
- |
2 - 3 |
413 |
63 |
View Full Product Details