Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- qkr58E-1
- Entrez Gene ID
-
37561
- Gene Name
- quaking related 58E-1
- Gene Aliases
-
CG3613, Dmel\CG3613, Dmel_CG3613, QKR58E-1, Qkr58E-1, qkr-58-E, qrk58E-1, qrk58E1
- Chromosome Location
-
Chr.2R: 22412556 - 22415518 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0022986
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079085.5 |
NP_523809.2 |
4 - 5 |
1165 |
66 |
GenBank mRNA |
AY118745.1 |
- |
4 - 5 |
1189 |
66 |
View Full Product Details