Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bgcn
- Entrez Gene ID
-
47873
- Gene Name
- benign gonial cell neoplasm
- Gene Aliases
-
BGCN, Bgcn, CG10330, CG10331, CG30170, Dmel\CG30170, Dmel_CG30170, PNBP, b(2)cgn, b(2)gcn, bcgn, ben(2)gcn, cg10331, fs(2)eo-C, fs(2)eo6, fs(2)eoQS2, gcn, pep
- Chromosome Location
-
Chr.2R: 23853270 - 23858921 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004581
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079108.3 |
NP_523832.2 |
1 - 2 |
1731 |
70 |
GenBank mRNA |
AF255662.1 |
- |
1 - 2 |
1601 |
70 |
View Full Product Details