Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nos
Entrez Gene ID
42297
Gene Name
nanos
Gene Aliases
CG5637, DRONANOS, Dmel\CG5637, Dmel_CG5637, Dronanos, NANOS, NOS, Nanos, Nos, l(3)07117, l(3)j3B6, l(3)j4B6, nNOS
Chromosome Location
Chr.3R: 19157238 - 19160202 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002962
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275794.1 NP_001262723.1 2 - 3 680 89
NM_057310.4 NP_476658.1 1 - 2 737 89
GenBank mRNA BT132708.1 - 1 - 2 713 89

View Full Product Details