Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx3
Entrez Gene ID
41551
Gene Name
Sorting nexin 3
Gene Aliases
CG6359, DmSNX3, Dmel\CG6359, Dmel_CG6359, Dsnx3, lincRNA.S6645, snx3
Chromosome Location
Chr.3R: 12628031 - 12630495 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038065
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141957.4 NP_650214.1 1 - 2 493 72
NM_169476.2 NP_731727.2 1 - 2 493 72
GenBank mRNA AY071431.1 - 1 - 2 483 72

View Full Product Details