Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atg8b
Entrez Gene ID
42132
Gene Name
Autophagy-related 8b
Gene Aliases
ATG8, ATG8b, Atg8, Atg8B, CG12334, Dmel\CG12334, Dmel_CG12334, atg8B, atg8b, dAtg8b
Chromosome Location
Chr.3R: 17724410 - 17725063 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038539
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316551.1 NP_001303480.1 1 - 1 549 76
NM_142392.3 NP_650649.1 1 - 1 530 76

View Full Product Details