Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir93a
- Entrez Gene ID
-
42471
- Gene Name
- Ionotropic receptor 93a
- Gene Aliases
-
CG17274, CG17275, CG42315, CT35907, DmelIR93a, Dmel\CG42315, Dmel_CG42315, IR93a, ir93a
- Chromosome Location
-
Chr.3R: 20851709 - 20859945 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259215
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142667.3 |
NP_650924.3 |
6 - 7 |
1067 |
62 |
View Full Product Details