Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kar
Entrez Gene ID
45883
Gene Name
karmoisin
Gene Aliases
CG12286, Dmel\CG12286, Dmel_CG12286
Chromosome Location
Chr.3R: 12682186 - 12689367 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001296
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202284.1 NP_001189213.1 4 - 5 1715 72
NM_143768.3 NP_652025.2 4 - 5 1715 72
GenBank mRNA BT021229.1 - 4 - 5 1717 72
BT099794.1 - 4 - 5 1715 72

View Full Product Details