Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SmydA-8
- Entrez Gene ID
-
31200
- Gene Name
- SET and MYND domain containing, arthropod-specific, member 8
- Gene Aliases
-
CG18033, CG32799, CG32800, CG33548, Dmel\CG33548, Dmel_CG33548, EG:103B4.4, EG:103B4.5, MSTA, Msta, msta
- Chromosome Location
-
Chr.X: 2205515 - 2213809 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0053548
View Full Product Details