Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
whip
Entrez Gene ID
5740782
Gene Name
whipple
Gene Aliases
CG2149, CG34218, Dmel\CG34218, Dmel_CG34218, NEST:bs34h06, NEST:bs36g01
Chromosome Location
Chr.2R: 8362359 - 8363218 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250827
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103760.2 NP_001097230.1 1 - 1 648 59

View Full Product Details