Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfand2a
Entrez Gene ID
393489
Gene Name
zinc finger, AN1-type domain 2A
Gene Aliases
zgc:66295, zgc:77923
Chromosome Location
Chr.3: 43373825 - 43393253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200517.2 NP_956811.1 8 - 9 835 74
XM_005164201.4 XP_005164258.1 9 - 10 1011 74
GenBank mRNA BC055589.1 - 9 - 10 959 74
BC065896.1 - 8 - 9 835 74

View Full Product Details