Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glipr1b
Entrez Gene ID
393547
Gene Name
GLI pathogenesis-related 1b
Gene Aliases
zgc:65887
Chromosome Location
Chr.18: 7345239 - 7355225 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200575.1 NP_956869.1 4 - 5 839 66
XM_005159001.4 XP_005159058.1 4 - 5 1041 66
GenBank mRNA BC056605.1 - 4 - 5 839 66

View Full Product Details