Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
derl1
Entrez Gene ID
406753
Gene Name
derlin 1
Gene Aliases
wu:fb09d07, wu:fb94d05, wu:fj92g12, zgc:55637
Chromosome Location
Chr.16: 25642936 - 25663844 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213444.2 NP_998609.2 1 - 2 338 87
GenBank mRNA BC045413.1 - 1 - 2 338 87
BC165504.1 - 1 - 2 190 87
EH551605.1 - 1 - 2 370 87

View Full Product Details