Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cryba1a
Entrez Gene ID
436683
Gene Name
crystallin, beta A1a
Gene Aliases
cryba1, zgc:92688
Chromosome Location
Chr.15: 28086694 - 28094349 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002410.1 NP_001002410.1 5 - 6 520 75
XM_005157649.4 XP_005157706.1 5 - 6 552 75
GenBank mRNA BC076170.1 - 5 - 6 520 75
DQ000462.1 - 5 - 6 520 75

View Full Product Details