Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pbrm1l
Entrez Gene ID
445141
Gene Name
polybromo 1, like
Gene Aliases
pb1, zgc:100857, zgc:113882
Chromosome Location
Chr.6: 251612 - 277433 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005168762.4 XP_005168819.1 15 - 16 1944 93
GenBank mRNA BC098878.1 - 4 - 5 290 93

View Full Product Details