Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pbrm1l
Entrez Gene ID
445141
Gene Name
polybromo 1, like
Gene Aliases
pb1, zgc:100857, zgc:113882
Chromosome Location
Chr.6: 251612 - 277433 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003535.2 NP_001003535.2 7 - 8 850 60
XM_005168762.4 XP_005168819.1 7 - 8 850 60
GenBank mRNA BC078299.1 - 7 - 8 854 60

View Full Product Details