Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cd63
Entrez Gene ID
321461
Gene Name
CD63 molecule
Gene Aliases
cd163, wu:fb16b08, zgc:65825, zgc:77709
Chromosome Location
Chr.23: 32066294 - 32085939 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199543.1 NP_955837.1 2 - 3 142 88
GenBank mRNA BC056698.1 - 2 - 3 142 88
BC064300.1 - 2 - 3 180 88

View Full Product Details