Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmed4
Entrez Gene ID
415224
Gene Name
transmembrane p24 trafficking protein 4
Gene Aliases
fc88h09, wu:fc88h09, zgc:86755
Chromosome Location
Chr.23: 33977724 - 33986743 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002134.1 NP_001002134.1 1 - 2 168 112
GenBank mRNA BC071427.1 - 1 - 2 168 112

View Full Product Details