Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
irf10
Entrez Gene ID
405815
Gene Name
interferon regulatory factor 10
Gene Aliases
zgc:76891
Chromosome Location
Chr.23: 641958 - 653333 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212879.1 NP_998044.1 6 - 7 896 156
XM_005161894.3 XP_005161951.1 6 - 7 925 156
GenBank mRNA BC066724.1 - 6 - 7 896 156
BC153458.1 - 6 - 7 687 156
BC153602.1 - 6 - 7 656 156
EU274625.1 - 5 - 6 156

View Full Product Details