Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OASL
Entrez Gene ID
100009702
Gene Name
2'-5'-oligoadenylate synthetase-like
Gene Aliases
-
Chromosome Location
Chr.8: 15623084 - 15639859 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081797.1 NP_001075266.1 2 - 3 484 75
XM_005612545.3 XP_005612602.1 - 75
GenBank mRNA AY463162.1 - 2 - 3 484 75

View Full Product Details