Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FERMT2
Entrez Gene ID
423589
Gene Name
fermitin family member 2
Gene Aliases
PLEKHC1
Chromosome Location
Chr.5: 58412614 - 58449902 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004936527.3 XP_004936584.2 14 - 15 1782 60
XM_025150869.1 XP_025006637.1 12 - 13 1776 60
GenBank mRNA CR389161.1 - 2 - 3 236 60

View Full Product Details