Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIDA
Entrez Gene ID
421335
Gene Name
axin interactor, dorsalization associated
Gene Aliases
C1orf80
Chromosome Location
Chr.3: 18103701 - 18128618 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277540.2 NP_001264469.1 1 - 2 238 97
GenBank mRNA BU113910.1 - 1 - 2 238 97
BX931410.1 - 1 - 2 187 97

View Full Product Details