Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DERL1
Entrez Gene ID
420350
Gene Name
derlin 1
Gene Aliases
derlin-1
Chromosome Location
Chr.2: 137901053 - 137916181 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006350.2 NP_001006350.1 4 - 5 427 82
GenBank mRNA AJ719425.1 - 4 - 5 429 82

View Full Product Details