Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TENM1
Entrez Gene ID
395668
Gene Name
teneurin transmembrane protein 1
Gene Aliases
ODZ1, TEN-1
Chromosome Location
Chr.4: 15205742 - 15529114 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204862.1 NP_990193.1 27 - 28 5609 69
XM_015278189.2 XP_015133675.1 28 - 29 6871 69
XM_015278190.2 XP_015133676.1 28 - 29 6868 69
XM_015278191.2 XP_015133677.1 27 - 28 6847 69
GenBank mRNA AJ238613.1 - 27 - 28 5609 69

View Full Product Details