Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM65
Entrez Gene ID
428387
Gene Name
transmembrane protein 65
Gene Aliases
-
Chromosome Location
Chr.2: 138399888 - 138435087 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031507.1 NP_001026678.1 2 - 3 293 104
XM_015283110.2 XP_015138596.1 4 - 5 901 104
XM_015283111.2 XP_015138597.1 3 - 4 830 104
GenBank mRNA AJ719948.1 - 2 - 3 293 104
CR391308.1 - 2 - 3 120 104

View Full Product Details