Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT5B
Entrez Gene ID
418154
Gene Name
Wnt family member 5B
Gene Aliases
-
Chromosome Location
Chr.1: 60852694 - 60918508 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037269.1 NP_001032346.1 2 - 3 332 58
XM_015285801.2 XP_015141287.1 3 - 4 712 58
GenBank mRNA AY753289.1 - 2 - 3 332 58

View Full Product Details