Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK9
Entrez Gene ID
417226
Gene Name
cyclin dependent kinase 9
Gene Aliases
-
Chromosome Location
Chr.17: 5268862 - 5275457 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006201.2 NP_001006201.1 4 - 5 468 77
GenBank mRNA AJ450765.1 - 4 - 5 469 77
AJ720053.1 - 4 - 5 445 77
DR425719.1 - 4 - 5 397 77

View Full Product Details