Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK9
Entrez Gene ID
417226
Gene Name
cyclin dependent kinase 9
Gene Aliases
-
Chromosome Location
Chr.17: 5268862 - 5275457 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006201.2 NP_001006201.1 6 - 7 797 102
GenBank mRNA AJ720053.1 - 6 - 7 774 102
DR425719.1 - 6 - 7 726 102

View Full Product Details