Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMADHC
Entrez Gene ID
424311
Gene Name
methylmalonic aciduria and homocystinuria, cblD type
Gene Aliases
C2orf25, CblD
Chromosome Location
Chr.7: 34920828 - 34933478 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008477.1 NP_001008477.1 2 - 3 341 66
GenBank mRNA AJ720042.1 - 2 - 3 341 66
BX936194.1 - 2 - 3 314 66

View Full Product Details