Gene Transcripts

Gene Symbol
HIST1H1T
Entrez Gene ID
3010
Gene Name
histone cluster 1, H1t
Gene Aliases
H1.6, H1FT, H1t, dJ221C16.2
Chromosome Location
Chr. 6: 26107412 - 26108136 on Build GRCh38
UniGene ID
Hs.533293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005323.3 NP_005314.2 1 357
GenBank mRNA BC069517.1 - 1 357
BC130350.1 - 1 350 40146381

View Full Product Details