Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr. 19: 45815442 - 45863290 on Build GRCh38
UniGene ID
Hs.515475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004819.2 NP_004810.2 18 2645
XM_005259286.1 XP_005259343.1 18 2598
XM_011527354.1 XP_011525656.1 19 2648
GenBank mRNA AB209024.1 - 18 4808
AB210010.1 - 18 2490
AK130602.1 - 9 784
AK300252.1 - 18 2499
BC006536.2 - 17 2352
BC006567.2 - 17 2352
BX649165.1 - 17 3551
U49240.1 - 13 2114
Y10931.1 - 18 2463

View Full Product Details