Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144772.1 | NP_001138244.1 | 20, 21 | 1770 | |
NM_003580.3 | NP_003571.2 | 20, 21 | 1830 | ||
GenBank mRNA | AK292679.1 | - | 20, 21 | 1710 | |
AK294009.1 | - | 20, 21 | 1770 | ||
AK313991.1 | - | 20, 21 | 1741 | ||
BC041124.1 | - | 20, 21 | 1812 | 5785460 | |
X96586.1 | - | 20, 21 | 1628 |