Gene Transcripts

Gene Symbol
INA
Entrez Gene ID
9118
Gene Name
internexin neuronal intermediate filament protein alpha
Gene Aliases
NEF5, NF-66, TXBP-1
Chromosome Location
Chr. 10: 103277163 - 103290351 on Build GRCh38
UniGene ID
Hs.500916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032727.3 NP_116116.1 1 908
GenBank mRNA AB209785.1 - 1 359
BC006359.2 - 1 908 4125949
S78296.1 - 1 892

View Full Product Details