Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286445.1 | NP_001273374.1 | 6 | 640 | |
NM_001286446.1 | NP_001273375.1 | 6 | 743 | ||
NM_001286447.1 | NP_001273376.1 | 6 | 840 | ||
NM_014722.3 | NP_055537.2 | 6 | 778 | ||
NM_015864.3 | NP_056948.2 | 6 | 840 | ||
XM_006715275.2 | XP_006715338.1 | 6 | 857 | ||
XM_006715277.1 | XP_006715340.1 | 6 | 778 | ||
XM_006715279.2 | XP_006715342.1 | 6 | 835 | ||
XM_006715281.3 | XP_006715344.1 | 6 | 857 | ||
XM_011515007.1 | XP_011513309.1 | 7 | 1366 | ||
XM_011515008.2 | XP_011513310.1 | 7 | 697 | ||
XM_011515009.1 | XP_011513311.1 | 6 | 569 | ||
XM_011515010.2 | XP_011513312.1 | 6 | 610 | ||
XM_011515012.1 | XP_011513314.1 | 6 | 857 | ||
XM_017011523.1 | XP_016867012.1 | 6 | 532 | ||
XM_017011524.1 | XP_016867013.1 | 6 | 786 | ||
XM_017011525.1 | XP_016867014.1 | 6 | 778 | ||
XM_017011526.1 | XP_016867015.1 | 6 | 778 | ||
XM_017011527.1 | XP_016867016.1 | 6 | 610 | ||
GenBank mRNA | AB002384.1 | - | 6 | 576 | |
AK299047.1 | - | 6 | 548 | ||
AK300138.1 | - | 6 | 788 | ||
AK300950.1 | - | 6 | 593 | ||
BC001232.1 | - | 6 | 830 | 3453890 | |
U49187.1 | - | 6 | 701 |