Gene Transcripts

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
Chromosome Location
Chr. 1: 200738905 - 200860707 on Build GRCh38
UniGene ID
Hs.23585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297707.1 NP_001284636.1 10 1450
NM_203459.2 NP_982284.1 9 1417
XM_017000799.1 XP_016856288.1 9 1843
GenBank mRNA AB029001.2 - 9 808
AK095640.1 - 9 1417
AK126821.1 - 9 822
BC125229.1 - 10 1229 40124603

View Full Product Details