Gene Transcripts

Gene Symbol
RAB30
Entrez Gene ID
27314
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 11: 82973133 - 83071923 on Build GRCh38
UniGene ID
Hs.40758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286059.1 NP_001272988.1 5 994
NM_001286060.1 NP_001272989.1 5 829
NM_001286061.1 NP_001272990.1 5 727
NM_014488.4 NP_055303.2 6 930
GenBank mRNA AF498956.1 - 4 588
AK291376.1 - 5 989
AK312323.1 - 5 735
BC014213.2 - 6 849 4763140
BX641138.1 - 5 774
CR542106.1 - 4 588
U57092.1 - 4 606

View Full Product Details