Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016322.3 | NP_057406.2 | 5 | 791 | |
GenBank mRNA | AF112206.1 | - | 5 | 399 | |
AF152463.1 | - | 4 | 335 | ||
AF203689.1 | - | 4 | 341 | ||
AF498949.1 | - | 4 | 335 | ||
AK023524.1 | - | 5 | 573 | ||
AK090889.1 | - | 5 | 590 | ||
AL162081.1 | - | 5 | 518 | ||
AY553875.1 | - | 4 | 335 | ||
BC006081.1 | - | 5 | 562 | 2963119 | |
CR457394.1 | - | 4 | 335 |