Gene Transcripts

Gene Symbol
ZFAND3
Entrez Gene ID
60685
Gene Name
zinc finger AN1-type containing 3
Gene Aliases
TEX27
Chromosome Location
Chr. 6: 37819496 - 38154624 on Build GRCh38
UniGene ID
Hs.36959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021943.2 NP_068762.1 3 570
XM_005249269.1 XP_005249326.1 3 605
XM_011514790.1 XP_011513092.1 3 605
XM_017011171.1 XP_016866660.1 2 543
GenBank mRNA AK023284.1 - 3 244
BC031481.2 - 3 186 4448968
BP305887.1 - 3 299

View Full Product Details