Gene Transcripts

Gene Symbol
TMEM9
Entrez Gene ID
252839
Gene Name
transmembrane protein 9
Gene Aliases
DERM4, TMEM9A
Chromosome Location
Chr. 1: 201134771 - 201171625 on Build GRCh38
UniGene ID
Hs.181444
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288564.1 NP_001275493.1 6 657
NM_001288565.1 NP_001275494.1 5 1160
NM_001288566.1 NP_001275495.1 6 758
NM_001288567.1 NP_001275496.1 6 972
NM_001288568.1 NP_001275497.1 6 810
NM_001288569.1 NP_001275498.1 6 671
NM_001288570.1 NP_001275499.1 6 684
NM_001288571.1 NP_001275500.1 6 750
NM_016456.4 NP_057540.1 6 675
NR_109920.1 7 902
NR_109921.1 5 1129
XM_011509385.1 XP_011507687.1 6 734
XM_011509386.2 XP_011507688.1 6 709
XM_011509387.2 XP_011507689.1 6 597
GenBank mRNA AB013909.1 - 5 528
AF151020.1 - 7 647
AK054883.1 - 6 630
AK075335.1 - 6 585
AK091726.1 - 5 1045
AK094588.1 - 2 1978
AK303843.1 - 6 696
AL136600.1 - 6 614
AY138587.1 - 6 586
AY359012.1 - 6 649
BC001106.2 - 6 599 3506442
CR457162.1 - 5 528
CR533467.1 - 5 534

View Full Product Details