Gene Transcripts
- Gene Symbol
- RAB4B-EGLN2
- Entrez Gene ID
- 100529264
- Gene Name
- RAB4B-EGLN2 readthrough (NMD candidate)
- Gene Aliases
- RERT-lncRNA
- Chromosome Location
- Chr. 19: 40778219 - 40808441 on Build GRCh38
- UniGene ID
- Hs.631539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037791.1 |
|
9 |
1933 |
|
GenBank mRNA |
AK291385.1 |
- |
9 |
1881 |
|
- Gene Symbol
- EGLN2
- Entrez Gene ID
- 112398
- Gene Name
- egl-9 family hypoxia inducible factor 2
- Gene Aliases
- EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
- Chromosome Location
- Chr. 19: 40799143 - 40808441 on Build GRCh38
- UniGene ID
- Hs.515417
- Species
- Homo sapiens
View Full Product Details