Gene Transcripts
- Gene Symbol
- TTTY21
- Entrez Gene ID
- 252953
- Gene Name
- testis-specific transcript, Y-linked 21 (non-protein coding)
- Gene Aliases
- NCRNA00146
- Chromosome Location
- Chr. Y: 9717653 - 9721296 on Build GRCh38
- UniGene ID
- Hs.522847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001535.1 |
|
1, 2 |
200 |
|
- Gene Symbol
- TTTY21B
- Entrez Gene ID
- 100101115
- Gene Name
- testis-specific transcript, Y-linked 21B (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. Y: 6443434 - 6447077 on Build GRCh38
- UniGene ID
- Hs.522847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003588.1 |
|
1, 2 |
200 |
|
View Full Product Details