Gene Transcripts

Gene Symbol
PDIA3P1
Entrez Gene ID
171423
Gene Name
protein disulfide isomerase family A member 3 pseudogene 1
Gene Aliases
ERp60, GRP58P, PDIA3P
Chromosome Location
Chr. 1: 147177851 - 147179949 on Build GRCh38
UniGene ID
Hs.658566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002305.1 1 1597
GenBank mRNA AK295985.1 - 1 1482
BI712496.1 - 1 500

View Full Product Details