Gene Transcripts

Gene Symbol
IZUMO1R
Entrez Gene ID
390243
Gene Name
IZUMO1 receptor, JUNO
Gene Aliases
FOLR4, Folbp3, JUNO
Chromosome Location
Chr. 11: 94305637 - 94307692 on Build GRCh38
UniGene ID
Hs.553758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199206.1 NP_001186135.1 3 380

View Full Product Details