Gene Transcripts

Gene Symbol
LRTM2
Entrez Gene ID
654429
Gene Name
leucine rich repeats and transmembrane domains 2
Gene Aliases
Chromosome Location
Chr. 12: 1820267 - 1836753 on Build GRCh38
UniGene ID
Hs.585579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039029.2 NP_001034118.1 4 1418
NM_001163925.1 NP_001157397.1 4 1454
NM_001163926.1 NP_001157398.1 4 1421
XM_011521015.1 XP_011519317.1 4 1639
XM_017019848.1 XP_016875337.1 2 826
GenBank mRNA AK095610.1 - 4 1156
AK124687.1 - 4 1025
BC129997.1 - 2 537
BC146894.1 - 4 1057 9056822
BC146910.1 - 4 1057 9056838
BC150567.1 - 2 537
BC150568.1 - 2 662 40127648

View Full Product Details