Gene Transcripts
- Gene Symbol
- IQCB1
- Entrez Gene ID
- 9657
- Gene Name
- IQ motif containing B1
- Gene Aliases
- NPHP5, PIQ, SLSN5
- Chromosome Location
- Chr. 3: 121769761 - 121835079 on Build GRCh38
- UniGene ID
- Hs.604110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001023570.3 |
NP_001018864.2
|
12, 13 |
1499 |
|
NM_001023571.3 |
NP_001018865.2
|
9, 10 |
1100 |
|
NM_001319107.1 |
NP_001306036.1
|
12, 13 |
1498 |
|
NR_134968.1 |
|
11, 12 |
1388 |
|
XM_005247911.3 |
XP_005247968.1
|
12, 13 |
1498 |
|
XM_005247912.2 |
XP_005247969.1
|
11, 12 |
1312 |
|
XM_011513335.2 |
XP_011511637.1
|
10, 11 |
1218 |
|
XM_017007537.1 |
XP_016863026.1
|
11, 12 |
1381 |
|
XM_017007538.1 |
XP_016863027.1
|
8, 9 |
1183 |
|
XM_017007539.1 |
XP_016863028.1
|
9, 10 |
1086 |
|
GenBank mRNA |
AB062481.1 |
- |
11, 12 |
1382 |
|
AY714228.1 |
- |
12, 13 |
1493 |
|
AY964667.1 |
- |
9, 10 |
1050 |
|
AY964668.1 |
- |
12, 13 |
1437 |
|
BC005806.2 |
- |
9, 10 |
1041 |
3834623 |
D25278.1 |
- |
12, 13 |
1440 |
|
View Full Product Details