Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11963454-11963965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5491021 c.1190T>C p.I397T Chr.1: 11963624-11963624 on Build GRCh38
6750247 c.1117C>T p.R373C Chr.1: 11963551-11963551 on Build GRCh38
5843259 c.1316G>A p.S439N Chr.1: 11963609-11963609 on Build GRCh38
5491022 c.1331T>C p.I444T Chr.1: 11963624-11963624 on Build GRCh38
4645046 c.1321C>T p.R441W Chr.1: 11963614-11963614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768317167 Chr.1: 11963568-11963568 on Build GRCh38
rs796446944 Chr.1: 11963771-11963772 on Build GRCh38
rs74949176 Chr.1: 11963598-11963598 on Build GRCh38
rs201322199 Chr.1: 11963496-11963496 on Build GRCh38
rs775997657 Chr.1: 11963819-11963819 on Build GRCh38
rs1056127956 Chr.1: 11963747-11963747 on Build GRCh38
rs148644646 Chr.1: 11963544-11963544 on Build GRCh38
rs574781992 Chr.1: 11963543-11963543 on Build GRCh38
rs745889627 Chr.1: 11963672-11963672 on Build GRCh38
rs1051492357 Chr.1: 11963846-11963846 on Build GRCh38
rs754748178 Chr.1: 11963564-11963564 on Build GRCh38
rs573420829 Chr.1: 11963764-11963764 on Build GRCh38
rs747725183 Chr.1: 11963649-11963649 on Build GRCh38
rs552366652 Chr.1: 11963886-11963886 on Build GRCh38
rs753853335 Chr.1: 11963631-11963631 on Build GRCh38
rs992966356 Chr.1: 11963708-11963709 on Build GRCh38
rs998877433 Chr.1: 11963926-11963926 on Build GRCh38
rs755934444 Chr.1: 11963521-11963521 on Build GRCh38
rs191004048 Chr.1: 11963871-11963871 on Build GRCh38
rs922826489 Chr.1: 11963832-11963833 on Build GRCh38
rs144439284 Chr.1: 11963616-11963616 on Build GRCh38
rs1051856481 Chr.1: 11963583-11963583 on Build GRCh38
rs749210400 Chr.1: 11963567-11963567 on Build GRCh38
rs761878111 Chr.1: 11963610-11963610 on Build GRCh38
rs879184052 Chr.1: 11963681-11963681 on Build GRCh38
rs376235626 Chr.1: 11963605-11963605 on Build GRCh38
rs374048093 Chr.1: 11963663-11963663 on Build GRCh38
rs942954664 Chr.1: 11963821-11963821 on Build GRCh38
rs998178222 Chr.1: 11963579-11963579 on Build GRCh38
rs776999916 Chr.1: 11963522-11963522 on Build GRCh38
rs907632411 Chr.1: 11963858-11963858 on Build GRCh38
rs371223839 Chr.1: 11963587-11963587 on Build GRCh38
rs372681138 Chr.1: 11963519-11963519 on Build GRCh38
rs369976401 Chr.1: 11963790-11963791 on Build GRCh38
rs371094317 Chr.1: 11963792-11963792 on Build GRCh38
rs549841216 Chr.1: 11963847-11963847 on Build GRCh38
rs543937291 Chr.1: 11963811-11963813 on Build GRCh38
rs1029629042 Chr.1: 11963933-11963933 on Build GRCh38
rs755978483 Chr.1: 11963628-11963628 on Build GRCh38
rs541901674 Chr.1: 11963529-11963529 on Build GRCh38
rs1030115518 Chr.1: 11963592-11963592 on Build GRCh38
rs1020789698 Chr.1: 11963771-11963773 on Build GRCh38
rs529709348 Chr.1: 11963839-11963839 on Build GRCh38
rs373520363 Chr.1: 11963599-11963599 on Build GRCh38
rs377497101 Chr.1: 11963549-11963549 on Build GRCh38
rs560674733 Chr.1: 11963834-11963834 on Build GRCh38
rs753127125 Chr.1: 11963537-11963537 on Build GRCh38
rs771685198 Chr.1: 11963586-11963586 on Build GRCh38
rs981042620 Chr.1: 11963646-11963646 on Build GRCh38
rs542516729 Chr.1: 11963779-11963779 on Build GRCh38
rs113555790 Chr.1: 11963531-11963531 on Build GRCh38
rs377018142 Chr.1: 11963690-11963690 on Build GRCh38
rs559916442 Chr.1: 11963731-11963731 on Build GRCh38
rs115762130 Chr.1: 11963938-11963938 on Build GRCh38
rs772734562 Chr.1: 11963791-11963791 on Build GRCh38
rs899107980 Chr.1: 11963625-11963625 on Build GRCh38
rs926169018 Chr.1: 11963812-11963812 on Build GRCh38
rs532268599 Chr.1: 11963874-11963874 on Build GRCh38
rs951362903 Chr.1: 11963934-11963934 on Build GRCh38
rs779852698 Chr.1: 11963487-11963487 on Build GRCh38
rs113474815 Chr.1: 11963530-11963530 on Build GRCh38
rs945090014 Chr.1: 11963780-11963780 on Build GRCh38
rs772509221 Chr.1: 11963517-11963519 on Build GRCh38
rs960392250 Chr.1: 11963701-11963701 on Build GRCh38
rs1041835691 Chr.1: 11963525-11963525 on Build GRCh38
rs1047089935 Chr.1: 11963626-11963626 on Build GRCh38
rs764179880 Chr.1: 11963607-11963607 on Build GRCh38
rs142423853 Chr.1: 11963910-11963910 on Build GRCh38
rs2230896 Chr.1: 11963575-11963575 on Build GRCh38
rs761521799 Chr.1: 11963505-11963505 on Build GRCh38
rs534570357 Chr.1: 11963918-11963918 on Build GRCh38
rs774190631 Chr.1: 11963502-11963502 on Build GRCh38
rs759962295 Chr.1: 11963524-11963524 on Build GRCh38
rs188970593 Chr.1: 11963480-11963480 on Build GRCh38
rs750301725 Chr.1: 11963551-11963551 on Build GRCh38
rs757830518 Chr.1: 11963650-11963650 on Build GRCh38
rs778574412 Chr.1: 11963645-11963645 on Build GRCh38
rs763409574 Chr.1: 11963606-11963606 on Build GRCh38
rs940338820 Chr.1: 11963865-11963867 on Build GRCh38
rs767839550 Chr.1: 11963615-11963615 on Build GRCh38
rs200131516 Chr.1: 11963574-11963574 on Build GRCh38
rs768259933 Chr.1: 11963501-11963501 on Build GRCh38
rs368762444 Chr.1: 11963506-11963506 on Build GRCh38
rs370358836 Chr.1: 11963639-11963639 on Build GRCh38
rs779676937 Chr.1: 11963629-11963629 on Build GRCh38
rs754151845 Chr.1: 11963823-11963825 on Build GRCh38

View Full Product Details