The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000261.1 | NP_000252.1 | 2 - 3 | 753 | 86 |
GenBank mRNA | AF001620.1 | - | 2 - 3 | 753 | 86 |
AK097310.1 | - | 3 - 4 | 701 | 86 | |
AK300924.1 | - | 3 - 4 | 629 | 86 | |
AK300943.1 | - | 3 - 4 | 754 | 86 | |
AK301297.1 | - | 3 - 4 | 674 | 86 | |
AK315443.1 | - | 2 - 3 | 803 | 86 | |
BC029261.1 | - | 2 - 3 | 791 | 86 | |
D88214.1 | - | 2 - 3 | 795 | 86 | |
FJ227302.1 | - | 2 - 3 | 753 | 86 | |
FJ237046.1 | - | 2 - 3 | 753 | 86 | |
FJ237047.1 | - | 2 - 3 | 753 | 86 | |
KU178210.1 | - | 2 - 3 | 731 | 86 | |
U85257.1 | - | 2 - 3 | 767 | 86 |