The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004334.2 | NP_004325.2 | 1 - 2 | 382 | 71 |
XM_005248185.1 | XP_005248242.1 | 1 - 2 | 382 | 71 | |
XM_005248186.1 | XP_005248243.1 | 1 - 2 | 382 | 71 | |
XM_011513878.2 | XP_011512180.1 | 1 - 2 | 382 | 71 | |
XM_011513879.1 | XP_011512181.1 | 1 - 2 | 382 | 71 | |
XM_017008565.1 | XP_016864054.1 | 1 - 2 | 382 | 71 | |
XM_017008566.1 | XP_016864055.1 | 1 - 2 | 382 | 71 | |
GenBank mRNA | AK312497.1 | - | 1 - 2 | 259 | 71 |
BC012095.1 | - | 1 - 2 | 257 | 71 | |
BT019502.1 | - | 1 - 2 | 187 | 71 | |
D21878.1 | - | 1 - 2 | 314 | 71 | |
DA382561.1 | - | 1 - 2 | 383 | 71 |