The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018945.3 | NP_061818.1 | 9 - 10 | 1109 | 57 |
XM_005266931.3 | XP_005266988.1 | 8 - 9 | 3958 | 57 | |
XM_011535755.2 | XP_011534057.1 | 10 - 11 | 938 | 57 | |
XM_017010741.1 | XP_016866230.1 | 8 - 9 | 1744 | 57 | |
GenBank mRNA | AB038040.1 | - | 9 - 10 | 1111 | 57 |
AB209990.1 | - | 8 - 9 | 1215 | 57 | |
AJ251860.1 | - | 9 - 10 | 1103 | 57 | |
AK290582.1 | - | 9 - 10 | 1142 | 57 | |
BC075082.2 | - | 9 - 10 | 832 | 57 | |
BC075083.2 | - | 9 - 10 | 836 | 57 | |
EF108327.1 | - | 8 - 9 | 1217 | 57 |